Learn More
Cayman Chemical Suramin (sodium salt), Purity: ≥98%, CAS: 129-46-4, 100 mg
SDP
Catalog No. 502951081
Encompass_Preferred
A polysulfonated naphthylurea with antiviral, antiparasitic, and anticancer activities; is negatively charged at physiological pH therefore exhibits polypharmacology; reduces Zika virus infectivity in Vero cells (IC50 = ~2.5-5 µg/ml); reduces the proliferation of bloodstream forms of T. brucei (MIC = 0.1 µM); reduces tumor volume in PDX mouse models of malignant mesothelioma at 60 mg/kg
Catalog No. 50-295-1081 Supplier Cayman Chemical Supplier No. 11126100 MG
A polysulfonated naphthylurea with antiviral, antiparasitic, and anticancer activities; is negatively charged at physiological pH therefore exhibits polypharmacology; reduces Zika virus infectivity in Vero cells (IC50 = ~2.5-5 µg/ml); reduces the proliferation of bloodstream forms of T. brucei (MIC = 0.1 µM); reduces tumor volume in PDX mouse models of malignant mesothelioma at 60 mg/kg