Learn More
Agilent Technologies SURE 2 SUPERCOMP CELLS
Catalog No. 50125018
Encompass
SURE 2 Supercompetent Cells. Increase stability of DNA containing inverted repeats or Z-DNA, produce high-quality miniprep DNA, contain F' episome allowing single-strand rescue and blue-white color screening, successfully clone unstable DNA
Catalog No. 50-125-018 Supplier Agilent Technologies Supplier No. 200152
SURE 2 Supercompetent Cells. Increase stability of DNA containing inverted repeats or Z-DNA, produce high-quality miniprep DNA, contain F' episome allowing single-strand rescue and blue-white color screening, successfully clone unstable DNA