Learn More
Sigma Aldrich Fine Chemicals Biosciences SuRE/CutTM Buffer H
Catalog No. 502900138
Encompass
The SuRE/Cut Buffer System consists of five optimized incubation buffers 10x concentrated for DNA restriction digest. Activity of all restriction enzymes has been determined in each buffer to select 100% activity or to calculate activity in double-digests.
Catalog No. 50-290-0138 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11417991001
The SuRE/Cut Buffer System consists of five optimized incubation buffers 10x concentrated for DNA restriction digest. Activity of all restriction enzymes has been determined in each buffer to select 100% activity or to calculate activity in double-digests.