Learn More
Agilent Technologies SURE ELECTROCOMP CELLS
Catalog No. 50125044
Encompass
SURE Electroporation-Competent Cells. For cloning DNA with secondary structures, > /= 1 x 1010 transformants/microgram. Clone unstable DNA, Increase stability of DNA containing inverted repeats or Z-DNA, Produce high-quality miniprep DNA
Catalog No. 50-125-044 Supplier Agilent Technologies Supplier No. 200227
SURE Electroporation-Competent Cells. For cloning DNA with secondary structures, > /= 1 x 1010 transformants/microgram. Clone unstable DNA, Increase stability of DNA containing inverted repeats or Z-DNA, Produce high-quality miniprep DNA