Learn More
Vici Precision Sampling, Inc. Sure Guard In-Line Filter Kit - High Pressure, Stainless Steel, 2 um, 3/pk
Supplier:  Vici Precision Sampling, Inc. JR0611SS2
Encompass_Preferred
This item has a minimum order quantity of 4 per supplier requirements. Frit Volume: 0.61 uL. 30 mL/min flow rate. The VICI Jour Sure-Guard disposable in-line filter offers an easy and inexpensive way to protect valuable columns against fines and particles. It is easily connected directly to any column, with an inlet for 1/16" OD tubes and 10-32 threads. The VICI Jour Sure-Guard can be changed in seconds without tools.
Catalog No. NC1371957