Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Sure-Grip™ EX Flammables Safety Can Storage Cabinets With Handles
SCI_ED

Catalog No. S28750 Shop All Justrite Safety Group Products
Change view
Click to view available options
Dimensions (D x W x H) Interior:
14.56 x 39.5 x 39.62 in.
30.34 x 39.5 x 60.5 in.
No. of Shelves:
1
2
Certifications/Compliance:
FM, NFPA, OSHA
FM, NFPA, OSHA, U (NFPA)
Door Style:
Manual-close
Self-close
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H) Interior No. of Shelves Certifications/Compliance Door Style
S28750 30.34 x 39.5 x 60.5 in. 2 FM, NFPA, OSHA, U (NFPA) Self-close
S28708 14.56 x 39.5 x 39.62 in. 1 FM, NFPA, OSHA, U (NFPA) Self-close
S28749 30.34 x 39.5 x 60.5 in. 2 FM, NFPA, OSHA Manual-close
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S28750 Supplier Justrite™ Supplier No. 899020/EMD

missing translation for 'validMainframeMsg'

Keep dangerous liquids safely organized and segregated. Justrite™ Sure-Grip™ EX Flammables Safety Can Storage Cabinets With Handles feature flush mounted handles, two keys for security and have the ability to be padlocked. Dual vents with flame arresters allow hazardous vapers to be piped away.

  • Sure-Grip EX flush-mounted door handle does not interfere with traffic; comes with two keys for security and can be padlocked (padlock not included)
  • Exclusive self-latching feature ensures that cabinet doors are safely closed without manually engaging door handle
  • Double-wall construction provides 1.5 in. (3.8cm) of insulating air space to help prevent spread of flames
  • High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away
  • Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self-latch engages critical three-point latching system for protection under fire conditions
  • Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
  • Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
  • Two-door models hinged left and right have self-closing mechanism that ensures proper closing sequence regardless of which door is released first
  • SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Adjustable leveling feet ensure solid positioning and alignment for tight door closure
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • Meet OSHA and NFPA requirements

Specifications

Product Type Can Storage Cabinet
Dimensions (D x W x H) Interior 30.34 x 39.5 x 60.5 in.
Load Bearing Capacity 90 gal.
Door Style Self-close
Height (Metric) Exterior 165.1 cm
Weight (Metric) 218 kg
Width (Metric) Interior 100.3 cm
Depth (Metric) Interior 77.1 cm
Height (Metric) Interior 153.7 cm
Depth (Metric) Exterior 86.4 cm
Width (Metric) Exterior 109 cm
No. of Doors 2
No. of Shelves 2
Depth (English) Interior 30.34 in.
Depth (English) Exterior 34 in.
Width (English) Exterior 43 in.
Certifications/Compliance FM, NFPA, OSHA, U (NFPA)
Height (English) Interior 60.5 in.
Includes U-Loc™ padlockable handle, Haz-Alert™ reflective trilingual warning labels, sturdy SpillSlope™ shelves, three-point Stainless Steel bullet self latching system and more!
Weight (English) 480 lb.
Height (English) Exterior 65 in.
Material Steel (CR)
Color Yellow
For Use With (Application) Flammable liquid storage and protection
Surface Coating Epoxy, Polyester
Width (English) Interior 39.5 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.