Learn More
Braintree Scientific Sure-Seal Induction Small Mouse Chamber 4"" x 4"" x 4""
SDP
Catalog No. 501954780
Encompass
Sure-Seal Induction Chambers. Initial anesthesia is performed in the induction chamber before animals are moved to the surgical bed for procedure. Positive seal gasket ensures there is no gas leakage. Simple locking snap latch. Gas is exhausted through an outlet port that is tube-connected to a charcoal filter canister. Can be oxygen-flushed before transferring animals out of chamber. Constructed of heavy-duty, clear acrylic plastic. Non-skid bottom **Sold for research purposes only and not for human use**
Catalog No. 50-195-4780 Supplier Braintree Scientific Supplier No. EZ177
Sure-Seal Induction Chambers. Initial anesthesia is performed in the induction chamber before animals are moved to the surgical bed for procedure. Positive seal gasket ensures there is no gas leakage. Simple locking snap latch. Gas is exhausted through an outlet port that is tube-connected to a charcoal filter canister. Can be oxygen-flushed before transferring animals out of chamber. Constructed of heavy-duty, clear acrylic plastic. Non-skid bottom **Sold for research purposes only and not for human use**