Learn More
World Precision Instrument Sure-Seal Large Mouse/Rat Induction Chamber
SDP
Catalog No. 502538841
Encompass
For use with EZ-Anesthesia
Seal gasket ensures no gas leakage
Simple locking snap latch
Gas exhaust outlet port connects to charcoal filter
Heavy-duty, acrylic
Non-skid bottom
Includes Filter Connection outlet hose, 1/2" ID
11.25L x 7W x 6H
Catalog No. 50-253-8841 Supplier World Precision Instrument Supplier No. EZ1785
For use with EZ-Anesthesia
Seal gasket ensures no gas leakage
Simple locking snap latch
Gas exhaust outlet port connects to charcoal filter
Heavy-duty, acrylic
Non-skid bottom
Includes Filter Connection outlet hose, 1/2" ID
11.25L x 7W x 6H