Learn More
World Precision Instrument Sure-Seal Mouse Induction Chamber
SDP
Catalog No. 502538842
Encompass
  • For use with EZ-Anesthesia
  • Seal gasket ensures no gas leakage
  • Simple locking snap latch
  • Gas exhaust outlet port connects to charcoal filter
  • Heavy-duty, acrylic
  • Non-skid bottom
  • Includes Filter Connection outlet hose, 1/2" ID
Catalog No. 50-253-8842 Supplier World Precision Instrument Supplier No. EZ177
  • For use with EZ-Anesthesia
  • Seal gasket ensures no gas leakage
  • Simple locking snap latch
  • Gas exhaust outlet port connects to charcoal filter
  • Heavy-duty, acrylic
  • Non-skid bottom
  • Includes Filter Connection outlet hose, 1/2" ID