Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Sure-Vue™ RPR Test Kits and Accessories, RPR Dispensing Needles
Shop All Fisherbrand Products

Click to view available options
Quantity:
10/Pk.
Description
For professional use only.
Specifications
Specifications
| CE Marker | Self-declared |
| Clia Complexity | Moderate (Serum, Plasma or Whole Blood), Waived (Other) |
| Type | Dispensing Needles |
| Disposable | Single-use |
| For Use With (Application) | Syphilis testing |
| Quantity | 10/Pk. |
| Form | Ready-to-Use (RTU) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction