Learn More
Fisher Healthcare™ Sure-Vue™ Disposable Oval Test Cards, 6-Well
SDP
Catalog No. 23038181 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
108/Pk.
Catalog No. Quantity
23-038-181 108/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-038-181
Add to Cart
Add to Cart
This item is not returnable. View return policy
Oval disposable test cards for use when testing for rheumatoid factor using the RF Slide Test Kit.
  • Each pack of disposable oval test cards contains 108 durable cards made of heavy cardstock
  • 6-well test cards with black ovals
  • Used for latex agglutination testing with the Fisher Healthcare™ Sure-Vue™ RF Slide Test Kit
Specifications
Color Black
CE Marker Self-declared
Clia Complexity Moderate (Serum, Plasma or Whole Blood), Waived (Other)
Type Test Cards
Disposable Single-use
No. of Wells 6 Wells
Product Line Sure-Vue
Quantity 108/Pk.
Sterility Non-sterile
Form Ready-to-Use (RTU)