Learn More
Fisher Healthcare™ Sure-Vue™ Disposable Rectangle Test Cards, 30-well
SDP
Catalog No. 23038182 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 Pk.
Catalog No. Quantity
23-038-182 25 Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-038-182
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pack of 30-well, rectangular disposable test cards for use when testing for rheumatoid factor using the RF Slide Test Kit.
Each pack of Disposable Rectangle Test Cards contains 25 durable cards made of heavy cardstock.
  • 30-well test cards with black rectangles
  • Used for latex agglutination testing with the Fisher Healthcare™ Sure-Vue™ RF Slide Test Kit
Specifications
Color Black
CE Marker Self-declared
Type Test Cards
Disposable Single-use
No. of Wells 30 Wells
Product Line Sure-Vue
Quantity 25 Pk.
Sterility Non-sterile