Learn More
Fisher Healthcare™ Sure-Vue™ Fecal Occult Blood Slide Test System
SDP
Catalog No. 23038033 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 Pack
1000 Pack
20 Pack
80 Pack
Type:
FOBT Developer
FOBT Developer (extra sen)
FOBT Single Slide (extra sen)
FOBT Single Slide Test
FOBT Take Home Kit
FOBT Take Home Kit (extra sen)
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-038033 20 Pack FOBT Developer
23-038036 80 Pack FOBT Take Home Kit (extra sen)
23-038031 100 Pack FOBT Single Slide Test
23-038032 80 Pack FOBT Take Home Kit
23-038040 1000 Pack FOBT Single Slide Test
23-038035 100 Pack FOBT Single Slide (extra sen)
23-038037 20 Pack FOBT Developer (extra sen)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 23-038033
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Available in traditional guaiac format for routine screening applications and extra-sensitive format for more stringent testing
  • Bottles of developer for use with Fecal Occult Blood Slide Test Systems.
  • 23-038033 is for use with normal slides
  • 23-038037 is for use with extra-sensitive slides
Specifications
Type FOBT Developer
For Use With (Application) Detection of fecal occult Blood
Includes 20 x 15mL Bottles of developer
Quantity 20 Pack