Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
The Fisher Healthcare™ Sure-Vue™ Mono Test is an agglutination test for the qualitative and semiquantitative detection of heterophil antibodies in human serum associated with infectious mononucleosis (IM).
- All testing materials included in each kit
- Easy to read results
- Can be used with serum or plasma
- Reactive and nonreactive controls with dropper tip vials included
- Storage requirement: 2 to 8°C
- Long shelf life
Specifications
Specifications
| CE Marker | Self-declared |
| Clia Complexity | Moderate |
| Detectable Analytes | Mononucleosis Heterophil Antibodies |
| Detection Method | Latex Agglutination |
| Sample Type | Serum, Plasma |
| Sensitivity | 1:28 Davidsohn Titer |
| Type | Mononucleosis Test |
| Formulation | Suspension of Horse Erythrocytes in Buffer with 0.1% Sodium Azide |
| Includes | Reagent, Controls, Test Cards, Disposable Stirrer Pipets |
| No. of Wells | 6 Wells |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
