Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Sure-Vue™ Mono Test Kit
SDP

Catalog No. p-7222909 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1000 Pk.
150 Pk.
50 Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-038-173 50 Pk.
23-038-174 150 Pk.
23-038-175 1000 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-038-173
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An agglutination test for the qualitative and semiquantitative detection of heterophil antibodies in human serum associated with infectious mononucleosis (IM).

The Fisher Healthcare™ Sure-Vue™ Mono Test is an agglutination test for the qualitative and semiquantitative detection of heterophil antibodies in human serum associated with infectious mononucleosis (IM).

  • All testing materials included in each kit
  • Easy to read results
  • Can be used with serum or plasma
  • Reactive and nonreactive controls with dropper tip vials included
  • Storage requirement: 2 to 8°C
  • Long shelf life

Specifications

CE Marker Self-declared
Clia Complexity Moderate
Detectable Analytes Mononucleosis Heterophil Antibodies
Detection Method Latex Agglutination
Sample Type Serum, Plasma
Sensitivity 1:28 Davidsohn Titer
Type Mononucleosis Test
Formulation Suspension of Horse Erythrocytes in Buffer with 0.1% Sodium Azide
Includes Reagent, Controls, Test Cards, Disposable Stirrer Pipets
No. of Wells 6 Wells
Product Line Sure-Vue
Quantity 50 Pk.
Specificity >95%
Sterility Non-sterile
Test Time 2 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.