Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Sure-Vue™ RF Slide Test Kit
SDP

Catalog No. 23038178 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 Pk.
25 Pk.
50 Pk.
500 Pk.
This item is not returnable. View return policy

Catalog No. 23-038-178

Edge

Due to product restrictions, please to purchase or view availability for this product.

This item is not returnable. View return policy

A slide agglutination assay for the qualitative and semiquantitative determination of rheumatoid factor (RF) in human serum.

The Fisher Healthcare™ Sure-Vue™ RF Slide Test is a slide agglutination assay for the qualitative and semiquantitative determination of rheumatoid factor (RF) in human serum.

  • Standardized to WHO reference
  • No initial dilution required
  • Reactive and nonreactive controls included
  • Long shelf life
  • Proven by CAP, AAB, and API surveys
  • Proudly made in the USA

Specifications

CE Marker Self-declared
Clia Complexity Moderate
Detectable Analytes Rheumatoid Factor (RF)
Detection Method Latex Agglutination
Sample Type Serum
Sensitivity 8.0 IU/mL
Type RF Test
Formulation Inert Latex Particles Coated with Gamma Globulins in 0.1% Sodium Azide
Includes Reagent, Dilution Buffer, Controls, Test Cards, Disposable Stirrer Pipets
No. of Wells 6 Wells
Product Line Sure-Vue™
Quantity 50 Pk.
Specificity >95%
Sterility Non-sterile
Test Time 2 min
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.