Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Sure-Vue™ RF Slide Test Kit
SDP

Catalog No. 23038180 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 Pk.
25 Pk.
50 Pk.
500 Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-038-180 500 Pk.
23-038-177 25 Pk.
23-038-178 50 Pk.
23-038-179 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-038-180
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A slide agglutination assay for the qualitative and semiquantitative determination of rheumatoid factor (RF) in human serum.

The Fisher Healthcare™ Sure-Vue™ RF Slide Test is a slide agglutination assay for the qualitative and semiquantitative determination of rheumatoid factor (RF) in human serum.

  • Standardized to WHO reference
  • No initial dilution required
  • Reactive and nonreactive controls included
  • Long shelf life
  • Proven by CAP, AAB, and API surveys
  • Proudly made in the USA

Specifications

CE Marker Self-declared
Clia Complexity Moderate
Detectable Analytes Rheumatoid Factor (RF)
Detection Method Latex Agglutination
Sample Type Serum
Sensitivity 8.0 IU/mL
Type RF Test
Formulation Inert Latex Particles Coated with Gamma Globulins in 0.1% Sodium Azide
Includes Reagent, Dilution Buffer, Controls, Test Cards, Disposable Stirrer Pipets
No. of Wells 6 Wells
Product Line Sure-Vue
Quantity 500 Pk.
Specificity >95%
Sterility Non-sterile
Test Time 2 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.