Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Sure-Vue™ RPR Test Kit
GSA_VASDP

Catalog No. 23038009 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 Tests
500 Tests
5000 Tests
Description:
RPR Test Kit
RPR Test Kit, 10-well
RPR Test Kit, 30-well
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Description
23-038009 100 Tests RPR Test Kit
23-038020 5000 Tests RPR Test Kit, 30-well
23-038021 5000 Tests RPR Test Kit, 10-well
23-038010 500 Tests RPR Test Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-038009
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Reagent, prediluted positive and negative controls, dispensing pipets, needle, plastic dispensing vial, disposable slides, and package insert

  • Antigen detects “reagin” present in serum of syphilitic patients
  • For in vitro diagnostic use

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Description RPR Test Kit
Detectable Analytes Reagin (Syphilis)
Type RPR Test Kit
For Use With (Application) For in-vitro diagnostic use
Includes Reagent, prediluted positive and negative control, dispensing pipets, needle, Plastic dispensing vial and disposable slides
Quantity 100 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.