Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Sure-Vue™ Rubella Test Kit
SDP

Catalog No. 23038184 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 Pk.
30 Pk.
500 Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-038-184 100 Pk.
23-038-183 30 Pk.
23-038-185 500 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-038-184
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A rapid latex particle agglutination test for the qualitative and semiquantitative determination of rubella virus antibodies in serum.

The Fisher Healthcare™ Sure-Vue™ Rubella Test is a rapid latex particle agglutination test for the qualitative and semiquantitative determination of rubella virus antibodies in serum.

  • Excellent reproducibility and outstanding accuracy
  • 3 levels of liquid with ready-to-use controls included
  • White latex agglutination test
  • Correlation with gold standard Hemagglutination Inhibition Test (HAI)
  • Long shelf life
  • Made in the USA

Specifications

CE Marker Self-declared
Clia Complexity Moderate
Detectable Analytes Rubella Virus Antibodies
Detection Method Latex Agglutination
Sample Type Serum
Sensitivity 1 to 2 IU/mL (Undiluted) or 10 to 20 IU/mL (Diluted)
Type Rubella Test
Formulation Inert Latex Particles Coated with Gamma Globulins in 0.1% Sodium Azide
Includes Reagent, Dilution Buffer, Controls, Test Cards, Disposable Stirrer Pipets
No. of Wells 9 Wells
Product Line Sure-Vue
Quantity 100 Pk.
Specificity >99%
Sterility Non-sterile
Test Time 8 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.