Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Sure-Vue™ Signature Rubella Test
SDP

Catalog No. 23038156 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 Tests
500 Test
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-038-156 500 Test
23-038-155 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-038-156
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Fisher Healthcare™ Sure-Vue™ Signature Rubella Test is a rapid latex particle agglutination test for the qualitative and semiquantitative determination of rubella virus antibodies in serum.

  • Excellent reproducibility and outstanding accuracy
  • Qualitative and semiquantitative results
  • 3 Levels of liquid, ready-to-use controls included
  • Results in 8 minutes
  • White latex agglutination test
  • Correlation with gold standard - Hemagglutination Inhibition Test (HAI)
  • Sensitivity: 10 to 20 IU/mL
  • Longest shelf life - up to 18 months from date of manufacture
  • Made in USA

Specifications

CE Marker Self-declared
Clia Complexity Moderate
Detectable Analytes Rubella Virus Antibodies
Detection Method Latex Agglutination
Sample Type Serum
Sensitivity 10 to 20 IU/mL
Type Rubella Latex Test
Formulation Inert Latex Particles Coated with Gamma Globulins, with 0.1% Sodium Azide
Includes Latex Reagent, Dilution Buffer, Reactive Control, Weak Reactive Control, Nonreactive Control, Disposable Test Cards, Disposible Stirrer Pipets
No. of Wells 9 Wells
Product Line Sure-Vue
Quantity 500 Test
Specificity >99%
Sterility Non-sterile
Test Time 8 min.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.