Learn More
Alkali Scientific SureAir PCR Workstation With U
SDP
Catalog No. 503029641
Encompass_Preferred
HEPA Filter and UV light reduce contamination Highly compact 22 x 16 footprint Included cable access port allows use of centrifuges vortexers and other instruments Included pipette holder and shelf provides storage for pipettes and tips Provides ISO 5 level clean conditions The Alkali SureAirPCR workstation is a compact convenient way to reduce contamination concerns and provide a clean room environment at a fraction of the space and cost The high-powered yet quiet internal fan produces laminar flow conditions without any installation into ductwork The included HEPA filter purifies air within the chamber meeting ISO 5 cleanliness standards and filters 99 97% of particles 0 3 m and larger The double-hinged front panel provides ample workspace for convenient hand access and can be opened fully to allow placement of equipment inside The SureAir is designed to keep your workspace clean easy-to-use and organized An included germicidal UVC light eliminates any stray DNA that may contam
Catalog No. 50-302-9641 Supplier Alkali Scientific Supplier No. RS5200
HEPA Filter and UV light reduce contamination Highly compact 22 x 16 footprint Included cable access port allows use of centrifuges vortexers and other instruments Included pipette holder and shelf provides storage for pipettes and tips Provides ISO 5 level clean conditions The Alkali SureAirPCR workstation is a compact convenient way to reduce contamination concerns and provide a clean room environment at a fraction of the space and cost The high-powered yet quiet internal fan produces laminar flow conditions without any installation into ductwork The included HEPA filter purifies air within the chamber meeting ISO 5 cleanliness standards and filters 99 97% of particles 0 3 m and larger The double-hinged front panel provides ample workspace for convenient hand access and can be opened fully to allow placement of equipment inside The SureAir is designed to keep your workspace clean easy-to-use and organized An included germicidal UVC light eliminates any stray DNA that may contam