Learn More
Seracare Life Sciences Inc SureBlue™ TMB 1-Component Microwell Peroxidase Substrate
Catalog No. 5067492
Encompass_Preferred
SureBlue TMB Microwell Substrate develops a deep blue color in the presence of peroxidase-labeled conjugates measurable at 650 nm and turns yellow when stopped by acidification and read at 450 nm. Recommended for ELISA and blotting applications with addition of membrane enhancer. Single component, ready to use format of 3,3', 5, 5' - Tetramethylbenzidine exibits exceptional stability and lot-to-lot consistency. No reagent preparation required, very convenient.
Requires storage 2-8 degrees Celsius.
This item is not returnable. View return policy
Catalog No. 50-674-92 Supplier Seracare Life Sciences Inc Supplier No. 51200076
SureBlue TMB Microwell Substrate develops a deep blue color in the presence of peroxidase-labeled conjugates measurable at 650 nm and turns yellow when stopped by acidification and read at 450 nm. Recommended for ELISA and blotting applications with addition of membrane enhancer. Single component, ready to use format of 3,3', 5, 5' - Tetramethylbenzidine exibits exceptional stability and lot-to-lot consistency. No reagent preparation required, very convenient.
Requires storage 2-8 degrees Celsius.