Learn More
Invitrogen™ SureCast™ Sealing Pads
Catalog No. HC1002
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 pads
Catalog No. Quantity
HC1002 2 pads
1 options
Catalog No. HC1002 Supplier Invitrogen™ Supplier No. HC1002
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Edge
Add to Cart
Leak-free, confident handcasting of your polyacrylamide gels with the Invitrogen™ SureCast™ Handcast System.
Leak-free, confident handcasting of your polyacrylamide gels with the Invitrogen™ SureCast™ Handcast System.
SureCast Sealing Pads are a replacement set of foam gaskets for use in the SureCast Handcast Station. Comes in a package of two (2) sealing pads.
  • Compatible only with the SureCast Handcast System
Recommended Storage
Room Temperature
Warranty and Services
Up to 12 months
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2 pads
Do you offer the SureCast casting reagents separately?
The SureCast Handcast System and reagents are available both as a package or as standalones.
Can I reuse SureCast Glass Plates?
Yes, SureCastGlass Plates can be reused.They are up to 22 times more durable than Bio-Rad glass plates.
Why should I store all SureCast reagents at room temperature
Having everything at room temperature means not having to thaw reagents, thus saving time in gel casting.
For Research Use Only. Not for use in diagnostic procedures.