Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SureCast™ Stacking Buffer

Catalog No. p-7184591
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 500 mL
5 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HC2112 2 x 500 mL
HC2115 5 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. HC2112 Supplier Invitrogen™ Supplier No. HC2112
Add to Cart
Add to Cart

These components of the SureCast Handcast System can be used to create polyacrylamide gels using SureCast Handcast Reagents as well as any other popular polyacrylamide gel casting reagents.

  • Pour polyacrylamide gels easily and confidently
  • For use with the SureCast vertical gel system
  • SureCast gels are designed for use in the Mini Gel Tank

Recommended Storage

Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 x 500 mL (1 L total) SureCast Stacking Buffer packs.

Store at room temperature.
Quantity 2 x 500 mL
Shipping Condition Room Temperature
Product Line SureCast
Product Type Stacking Buffer
Do you offer the SureCast casting reagents separately?

The SureCast Handcast System and reagents are available both as a package or as standalones.

Can I reuse SureCast Glass Plates?

Yes, SureCastGlass Plates can be reused.They are up to 22 times more durable than Bio-Rad glass plates.

Why should I store all SureCast reagents at room temperature

Having everything at room temperature means not having to thaw reagents, thus saving time in gel casting.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.