Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SureClick™ Antibody Labeling Kit

Catalog No. SC002
Encompass_Preferred
Change view
Click to view available options
Quantity:
3 x 1 mg
3 x 100 μg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SC002 3 x 1 mg
SC001 3 x 100 μg
SC003 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SC002 Supplier Invitrogen™ Supplier No. SC002
Only null left
Add to Cart
Edge
Add to Cart

Invitrogen SureClick Antibody Labeling Kits enable fast, precise, and reproducible conjugation of human or rabbit IgG antibodies using trusted click chemistry. Designed for spatial biology researchers and biotherapeutic developers, SureClick kits attach an average of 2 payloads per antibody at the Fc region.

Covalent, Fc-targeted antibody conjugation in just two hours

Invitrogen SureClick Antibody Labeling Kits enable fast, precise, and reproducible conjugation of human or rabbit IgG antibodies using trusted click chemistry. Designed for spatial biology researchers and biotherapeutic developers, SureClick kits attach an average of 2 payloads per antibody at the Fc region, preserving antigen binding and Fc receptor (FcR) affinity.

SureClick is an exceptional solution for attaching oligonucleotides to antibodies, making it ideal for spatial biology workflows, immuno-PCR, and next-generation antibody-based detection platforms that require precise and reproducible antibody–oligo conjugation.

Achieve consistent, covalent antibody labeling with a simplified workflow amenable to automation.

What is SureClick antibody labeling?

SureClick is a site-specific antibody conjugation technology that covalently attaches payloads—including fluorophores, functional tags, and oligonucleotides—to the Fc region of IgG antibodies using click chemistry. Unlike conventional random labeling approaches, SureClick:

  • Targets the Fc domain
  • Preserves Fab antigen-binding regions
  • Maintains Fc glycosylation integrity
  • Enables reproducible degree of labeling (DOL)
  • Supports consistent antibody–oligonucleotide conjugation

Key features and benefits:

Precise, reproducible labeling

  • Site-specific Fc conjugation
  • Consistent attachment of ~2 payloads per antibody
  • Reduced heterogeneity compared to conventional methods

Rapid two-hour workflow

  • ~10 minutes hands-on time
  • 90-minute protocol
  • Simple protocol using a single Fc-targeting component

Covalent and stable conjugation

  • Trusted click chemistry
  • Robust linkage for downstream assays, spatial biology, and antibody–oligo workflows

Preserves antibody function

  • Confidence that antigen binding is unchanged
  • Maintains Fc receptor interactions

Automation-ready

  • Spin-column purification
  • Minimal components
  • Suitable for high-throughput workflows

How SureClick antibody labeling works

  1. Add SureClick Fc-targeting reagent to your IgG antibody
  2. Purify using included Zeba™ spin columns
  3. Add click chemistry payload of choice (e.g., fluorophores, oligonucleotides, functional tags)

The result is a site-specific, covalently labeled antibody with consistent performance, including highly controlled antibody–oligonucleotide conjugates.

Applications

  • Spatial biology
  • Antibody–oligonucleotide conjugation
  • Multiplex biomarker detection
  • Antibody-drug conjugate (ADC) development
  • Biotherapeutic characterization
  • Flow cytometry
  • Internalization studies
  • Biomarker validation
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody Production and Labeling
Product Type Antibody Labeling Kit
Quantity 3 x 1 mg
Sufficient For Labeling 3 Antibodies
Content And Storage SureClick™ Fc Targeting Reagent
2°C to 8°C
For Use With (Equipment) Benchtop Centrifuge
Labeling Method Click Chemistry
Labeling Scale Approx. 2°
Shipping Condition Wet Ice
Product Line SureClick™
Chemical Reactivity Azide
Reactive Moiety Azide
Solubility Aqueous Buffer
Label or Dye Azide
Spacer Mid-length
Format Labeling Kit
Labeling Target Antibodies
Species Human or Rabbit
Incubation Time 70 to 90 min.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.