Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ SureFit™ Ergo-F Pipet Tips

Catalog No. 02681483 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
10 to 200 μL
Sterility:
Non-sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility
02-681-483 10 to 200 μL Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-681-483

missing translation for 'validMainframeMsg'

Designed specifically for use with Fisherbrand pipetters

  • Available in three capacities, with or without aerosol barrier
  • Unique wall design provides an extra-strong seal specific to Fisherbrand pipetters
  • Require minimal ejection force
  • Certified by independent laboratory - Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 to 200 μL
Filtered Non-filtered
Sterility Non-sterile
Format Refill
For Use With (Equipment) Thermo Scientific Finnpipette
Product Line SureFit Ergo-F
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.