Learn More
Alkali Scientific SureFrame™ Two-Component PCR Plate, 384-Well, Fully Skirted PC Frame, 35μL Clear Wells, 50/pk
SDP
Catalog No. 503027665
Encompass_Preferred
SureFrame™ plates are made with a special two-material process that creates an ultra-rigid polycarbonate frame while maintaining the favorable thermal transfer characteristics of thin-walled polypropylene tube wells. This ultra-rigid frame is ideal for all automated platforms and plate grippers and increases the uniformity of your experiments. Traditional single-material plates have been shown to warp inside thermal cyclers. This deformation of the plates causes increased evaporation in the outer wells of the plate, leading to inconsistent amplification across wells. SureFrame’s polycarbonate frame eliminates warping, for completely uniform results.   Two material design for increased rigidity Ideal for automated platforms Reduces evaporation Certified for PCR
Catalog No. 50-302-7665 Supplier Alkali Scientific Supplier No. PCR9384FSC
SureFrame™ plates are made with a special two-material process that creates an ultra-rigid polycarbonate frame while maintaining the favorable thermal transfer characteristics of thin-walled polypropylene tube wells. This ultra-rigid frame is ideal for all automated platforms and plate grippers and increases the uniformity of your experiments. Traditional single-material plates have been shown to warp inside thermal cyclers. This deformation of the plates causes increased evaporation in the outer wells of the plate, leading to inconsistent amplification across wells. SureFrame’s polycarbonate frame eliminates warping, for completely uniform results.   Two material design for increased rigidity Ideal for automated platforms Reduces evaporation Certified for PCR