Learn More
Cayman Chemical Surelght PRIdInInCLOroph 100ea
SDP
Catalog No. 502951922
Encompass_Preferred
SureLight Peridinin-Chlorophyll Protein Complex (PerCP)Excitation max.: 483 nmEmission max.: 676 nmUses: Flow cytometry/Fluorescence-activated cell sorting
Catalog No. 50-295-1922 Supplier Cayman Chemical Supplier No. 11271100 EA
SureLight Peridinin-Chlorophyll Protein Complex (PerCP)Excitation max.: 483 nmEmission max.: 676 nmUses: Flow cytometry/Fluorescence-activated cell sorting