Learn More
Cayman Chemical Surelght PRIdInInCLOroph 500ea
SDP
Catalog No. 502951499
Encompass_Preferred
SureLight Peridinin-Chlorophyll Protein Complex (PerCP)Excitation max.: 483 nmEmission max.: 676 nmUses: Flow cytometry/Fluorescence-activated cell sorting
Catalog No. 50-295-1499 Supplier Cayman Chemical Supplier No. 11271500 EA
Add to Cart
Add to Cart
Request a Quote
SureLight Peridinin-Chlorophyll Protein Complex (PerCP)Excitation max.: 483 nmEmission max.: 676 nmUses: Flow cytometry/Fluorescence-activated cell sorting