Learn More
Cayman Chemical SureLIght PRIdInInCLOroph 50ea
SDP
Catalog No. 502951155
Encompass_Preferred
SureLight Peridinin-Chlorophyll Protein Complex (PerCP)Excitation max.: 483 nmEmission max.: 676 nmUses: Flow cytometry/Fluorescence-activated cell sorting
Catalog No. 50-295-1155 Supplier Cayman Chemical Supplier No. 1127150 EA
SureLight Peridinin-Chlorophyll Protein Complex (PerCP)Excitation max.: 483 nmEmission max.: 676 nmUses: Flow cytometry/Fluorescence-activated cell sorting