Learn More
Cayman Chemical SureLIght488 NHS ester 1mg
SDP
Catalog No. 502939291
Encompass_Preferred
SureLight™ 488 is a bright green fluorescent dye with excitation suited to the 488 nm laser line, fluorescent microscopy, and flow cytometry. The NHS ester version is an efficient amine reactive probe that can be used to label proteins, antibodies, haptens, and other primary amine containing molecules.[Columbia Biosciences Product No. D44-100]
Catalog No. 50-293-9291 Supplier Cayman Chemical Supplier No. 270911 MG
SureLight™ 488 is a bright green fluorescent dye with excitation suited to the 488 nm laser line, fluorescent microscopy, and flow cytometry. The NHS ester version is an efficient amine reactive probe that can be used to label proteins, antibodies, haptens, and other primary amine containing molecules.[Columbia Biosciences Product No. D44-100]