Learn More
Cayman Chemical SureLIghtAlophycocynInA 100mg
SDP
Catalog No. 502965484
Encompass_Preferred
An estrogen metabolite that acts as a substrate of MRP2 (Km 75 UM) and through MRP2-mediated transport functions as a cholestatic agent decreasing bile flow
Catalog No. 50-296-5484 Supplier Cayman Chemical Supplier No. 16583100 MG
An estrogen metabolite that acts as a substrate of MRP2 (Km 75 UM) and through MRP2-mediated transport functions as a cholestatic agent decreasing bile flow