Learn More
Cayman Chemical SureLIghtAlophycocynInA 10mg
SDP
Catalog No. 502966395
Encompass_Preferred
Immunogen Synthetic peptide from the C-terminal region of human nNOS Host Rabbit Cross Reactivity (−) eNOS and iNOS Species Reactivity ( ) Human and rat Applications ICC IHC and WB
Catalog No. 50-296-6395 Supplier Cayman Chemical Supplier No. 1658310 MG
Immunogen Synthetic peptide from the C-terminal region of human nNOS Host Rabbit Cross Reactivity (−) eNOS and iNOS Species Reactivity ( ) Human and rat Applications ICC IHC and WB