Learn More
Cayman Chemical SureLIghtAlophycocynInA 50mg
SDP
Catalog No. 502965368
Encompass_Preferred
Immunogen Synthetic peptide from the C-terminal region of human eNOS Host Rabbit Species Reactivity ( ) bovine and human Cross Reactivity (−) iNOS and nNOS Application WB
Catalog No. 50-296-5368 Supplier Cayman Chemical Supplier No. 1658350 MG
Immunogen Synthetic peptide from the C-terminal region of human eNOS Host Rabbit Species Reactivity ( ) bovine and human Cross Reactivity (−) iNOS and nNOS Application WB