Learn More
Cayman Chemical SureLIghtBPhycoerythrInB 100mg
SDP
Catalog No. 502966111
Encompass_Preferred
Because of its high quantum yield, B-PE is considered the world’s brightest fluorophore. It is compatible with commonly available lasers and gives exceptional results in flow cytometry, Luminex and immunofluorescent staining. B-PE is also less “sticky” than common synthetic fluorophores and therefore gives less background interference. Uses: Flow cytometry, Fluorescence-activated cell sorting, Custom Antibody labeling, Luminex, and T Cell labeling
Catalog No. 50-296-6111 Supplier Cayman Chemical Supplier No. 16632100 MG
Because of its high quantum yield, B-PE is considered the world’s brightest fluorophore. It is compatible with commonly available lasers and gives exceptional results in flow cytometry, Luminex and immunofluorescent staining. B-PE is also less “sticky” than common synthetic fluorophores and therefore gives less background interference. Uses: Flow cytometry, Fluorescence-activated cell sorting, Custom Antibody labeling, Luminex, and T Cell labeling