Learn More
Cayman Chemical SureLIghtGlow FIrefly Luc
SDP
Catalog No. 502911010
Encompass_Preferred
A synthetic peptide substrate can be used as a substrate to quantify calcineurin activity
Catalog No. 50-291-1010 Supplier Cayman Chemical Supplier No. 34972100 TEST
A synthetic peptide substrate can be used as a substrate to quantify calcineurin activity