Learn More
Cayman Chemical SureLIghtPE-695 100mg
SDP
Catalog No. 502979771
Encompass_Preferred
An allosteric reversible inhibitor of PAR1 on platelets preventing surface expression of P-selectin induced by the peptide SFLLRN with an IC50 value of 0.26 UM also blocks platelet activation induced by thrombin but not by PMA U-46619 or collagen
Catalog No. 50-297-9771 Supplier Cayman Chemical Supplier No. 17465100 MG
An allosteric reversible inhibitor of PAR1 on platelets preventing surface expression of P-selectin induced by the peptide SFLLRN with an IC50 value of 0.26 UM also blocks platelet activation induced by thrombin but not by PMA U-46619 or collagen