Learn More
Cayman Chemical SureLIghtPE-695 5mg
SDP
Catalog No. 502965539
Encompass_Preferred
A highly potent reversible inhibitor of the EGFR (Ki 5.2 pM IC50 29 pM) rapidly suppresses autophosphorylation of EGFR in fibroblasts and human epidermoid carcinoma cells
Catalog No. 50-296-5539 Supplier Cayman Chemical Supplier No. 174655 MG
A highly potent reversible inhibitor of the EGFR (Ki 5.2 pM IC50 29 pM) rapidly suppresses autophosphorylation of EGFR in fibroblasts and human epidermoid carcinoma cells