Learn More
Cayman Chemical SureLIghtPerCP CynIn 5 5 100mg
SDP
Catalog No. 502910658
Encompass_Preferred
SureLight™ PerCP Cyanine 5.5 is a fluorescent probe used in protein detection Excitation max. λ: 496 nm Emission max. λ: 517 nm Uses: Custom antibody labeling, fluorescence-activated cell sorting, flow cytometry, Luminex, and T cell labeling
Catalog No. 50-291-0658 Supplier Cayman Chemical Supplier No. 34968100 MG
SureLight™ PerCP Cyanine 5.5 is a fluorescent probe used in protein detection Excitation max. λ: 496 nm Emission max. λ: 517 nm Uses: Custom antibody labeling, fluorescence-activated cell sorting, flow cytometry, Luminex, and T cell labeling