Learn More
Cayman Chemical SureLIghtR-PhycocynInRPC 100mg
SDP
Catalog No. 502909629
Encompass_Preferred
SureLight™ R-Phycocyanin comes from a red algaUses: Custom antibody labeling, fluorescence-activated cell sorting, and flow cytometry
Catalog No. 50-290-9629 Supplier Cayman Chemical Supplier No. 35481100 MG
SureLight™ R-Phycocyanin comes from a red algaUses: Custom antibody labeling, fluorescence-activated cell sorting, and flow cytometry