Learn More
Seracare Life Sciences Inc SURELINK ACTIVATED HRP
Catalog No. 5067593
Encompass_Preferred
SureLINK Activated HRP is a pre-activated form of horseradish peroxidase that makes it easier than ever to label your proteins or antibodies with HRP! Each vial contains 0.3 mg of lyophilized HRP that is ideal for conjugating between 0.05 mg to 0.1 mg of protein depending on its molecular weight. A comprehensive, step-by-step protocol offers guidance on what ratios to use for your specific situation.SureLINK Activated HRP can be used with your own conjugation protocol or follow the easy 2-step SureLINK HRP protocol and have HRP-conjugate ready to use in just 90 minutes.
This item is not returnable. View return policy
Catalog No. 50-675-93 Supplier Seracare Life Sciences Inc Supplier No. 56200023
SureLINK Activated HRP is a pre-activated form of horseradish peroxidase that makes it easier than ever to label your proteins or antibodies with HRP! Each vial contains 0.3 mg of lyophilized HRP that is ideal for conjugating between 0.05 mg to 0.1 mg of protein depending on its molecular weight. A comprehensive, step-by-step protocol offers guidance on what ratios to use for your specific situation.SureLINK Activated HRP can be used with your own conjugation protocol or follow the easy 2-step SureLINK HRP protocol and have HRP-conjugate ready to use in just 90 minutes.