Learn More
Seracare Life Sciences Inc SURELINK MODIFIED AP
Supplier:  Seracare Life Sciences Inc 56200028
Encompass_Preferred
SureLINK Modified AP is a modified form of alkaline phosphatase (AP) that is ideal for conjugating AP to monoclonal antibodies or other proteins! AP has been modified with the linker succinimidyl 4-hydrazinonicotinate (SANH). Based on unique conjugation chemistry, the Modified AP is part of a two-step coupling reaction. The conjugation reaction takes approximately 2 hours to complete. SureLINK Modified AP, 0.2 mg is ideal for conjugating 0.05 mg to 0.1 mg of antibody or protein. This product must be used with SFB-modified biolmolecules. It is simple and fast to attach to your antibody or protein.
Catalog No. 50-675-97