Learn More
LICORbio Xcell and Surelock Tank Adaptor (2 adaptors)
Catalog No. 502773680
Encompass_Preferred
Use these adapters to run Odyssey Mini Gels in Invitrogen XCell SureLock and other compatible electrophoresis tanks.These adaptors are not required for Bio-Rad Mini-PROTEAN systems. Invitrogen and XCell SureLock are trademarks or registered trademark of Thermo Fisher Scientific, Inc. in the US and other countries. Mini-PROTEAN is a trademark or registered trademark of Bio-Rad Laboratories, Inc. in the US and other countries.
Catalog No. 50-277-3680 Supplier LICORbio Supplier No. 90021168
Use these adapters to run Odyssey Mini Gels in Invitrogen XCell SureLock and other compatible electrophoresis tanks.These adaptors are not required for Bio-Rad Mini-PROTEAN systems. Invitrogen and XCell SureLock are trademarks or registered trademark of Thermo Fisher Scientific, Inc. in the US and other countries. Mini-PROTEAN is a trademark or registered trademark of Bio-Rad Laboratories, Inc. in the US and other countries.