Learn More
Phenomenex Inc Sure-Lok High Pressure PEEK 1-Pc Nut
Catalog No. 502401110 Shop All Phenomenex Products
Encompass_Preferred
Sure-Lok High Pressure PEEK 1-Pc Nut 10-32, For 1/16in Tubing, 12,000psi
Catalog No. 50-240-1110 Supplier Phenomenex Inc Supplier No. AQ08502
Sure-Lok High Pressure PEEK 1-Pc Nut 10-32, For 1/16in Tubing, 12,000psi