Learn More
Phenomenex Inc Sure-Lok High Pressure PEEK 1-Pc Nut - 10/Pk
Catalog No. 502398881 Shop All Phenomenex Products
Encompass_Preferred
Sure-Lok High Pressure PEEK 1-Pc Nut - 10/Pk 10-32, For 1/16in Tubing, 12,000psi
Catalog No. 50-239-8881 Supplier Phenomenex Inc Supplier No. AQ08503
Sure-Lok High Pressure PEEK 1-Pc Nut - 10/Pk 10-32, For 1/16in Tubing, 12,000psi