Learn More
Phenomenex Inc for 1/16in OD Tubing, 10-32 Thread, 19,000psi
Catalog No. 502397763 Shop All Phenomenex Products
Encompass_Preferred
Sure-Lok Nut, High Pressure, PEEK AQ0-8504
Catalog No. 50-239-7763 Supplier Phenomenex Inc Supplier No. AQ08504
Sure-Lok Nut, High Pressure, PEEK AQ0-8504