Learn More
Genscript Corporation SurePAGE™, Bis-Tris, 10x8, 4-20%, 15 wells
Catalog No. NC1629649
Encompass
GenScript SurePAGE, Bis-Tris, 10 cm x 8 cm gels are high-performance precast mini polyacrylamide gels with a special design that allows large sample loading volumes. The unique formulation of the gel and cassette design enables superior band resolution and significantly improved band evenness. SurePAGE gels are cast in a neutral pH buffer that minimizes polyacrylamide hydrolysis, increases gel stability and minimizes protein modification.
Catalog No. NC1629649 Supplier Genscript Corporation Supplier No. M00657
GenScript SurePAGE, Bis-Tris, 10 cm x 8 cm gels are high-performance precast mini polyacrylamide gels with a special design that allows large sample loading volumes. The unique formulation of the gel and cassette design enables superior band resolution and significantly improved band evenness. SurePAGE gels are cast in a neutral pH buffer that minimizes polyacrylamide hydrolysis, increases gel stability and minimizes protein modification.