Learn More
Genscript Corporation SurePAGE Bis-Tris 10x8 8-1
Catalog No. 5000057334
Encompass
GenScript SurePAGE Bis-Tris 10 cm x 8 cm gels are high-performance precast mini polyacrylamide gels with a special design that allows large sample loading volumes The unique formulation of the gel and cassette design enables superior band resolution and significantly improved band evenness SurePAGE gels are cast in a neutral pH buffer that minimizes polyacrylamide hydrolysis increases gel stability and minimizes protein modification br/ SurePAGE gels guarantee excellent batch-to-batch consistency and a reliable protein migration pattern With specially formulated Tris-MOPS running buffer proteins can be separated quickly and efficiently for subsequent detection by staining or Western blotting br/ br/ Bis-Tris 8-16% 10 wells br/ Cassette size 100 x 84 5 x 4 7 mm (L x W x T) br/ Gel size 80 x 70 x 1 mm (L x W x T)
Catalog No. 50-000-57334 Supplier Genscript Corporation Supplier No. M00658
GenScript SurePAGE Bis-Tris 10 cm x 8 cm gels are high-performance precast mini polyacrylamide gels with a special design that allows large sample loading volumes The unique formulation of the gel and cassette design enables superior band resolution and significantly improved band evenness SurePAGE gels are cast in a neutral pH buffer that minimizes polyacrylamide hydrolysis increases gel stability and minimizes protein modification br/ SurePAGE gels guarantee excellent batch-to-batch consistency and a reliable protein migration pattern With specially formulated Tris-MOPS running buffer proteins can be separated quickly and efficiently for subsequent detection by staining or Western blotting br/ br/ Bis-Tris 8-16% 10 wells br/ Cassette size 100 x 84 5 x 4 7 mm (L x W x T) br/ Gel size 80 x 70 x 1 mm (L x W x T)