Learn More
Thermo Scientific™ SureQuant™ AKT Pathway (Phospho) Absolute Quantitation Module
Description
- Complete - contains validated AQUA HeavyPeptide and LightPeptide mixtures, diluent, matrix, system suitability standard including optimized acquisition and analysis methods
- Validated - peptides rigorously tested for successful quantitation of each target peptide
- Multiplex - able to quantitate phosphorylated AKT, PTEN, IRS1, IGF1-R, GSK3a, GSK3b, PRAS40, mTOR, p70S6K, and TSC2 in a single reaction
Specifications
Specifications
| Detection Method | Mass Spectrometry |
| For Use With (Equipment) | Thermo Scientific™ Q Exactive Mass Spectrometer, Mass Spectrometer |
| Shipping Condition | Approved for shipment on Wet or Dry Ice |
| Workflow Step | Target Selection |
| Product Line | SureQuant |
| Quantity | 10 reactions kit |
| For Use With (Application) | Mass Spectrometry |
| Type | Quantitation Module |
| Final Product Type | Peptides |
| Starting Material | Protein Samples |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.