Learn More
Alkali Scientific ScrewSeal™ Screw Cap Microcentrifuge Tube
SDP
Catalog No. 502389226
Encompass_Preferred
SureSeal™ 1.5mL (45.5 x 10.5mm), with loop caps, sterile, with O-ring, conical bottom, non-graduated, caps assembled, 100 per bag.
Catalog No. 50-238-9226 Supplier Alkali Scientific Supplier No. MT3150SL
SureSeal™ 1.5mL (45.5 x 10.5mm), with loop caps, sterile, with O-ring, conical bottom, non-graduated, caps assembled, 100 per bag.